missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytohesin 2 Mouse anti-Human, Unconjugated, Clone: 3A7A1, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Cytohesin 2 |
---|---|
Clone | 3A7A1 |
Concentration | 1000 μg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16849685
|
Proteintech
67185-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16839685
|
Proteintech
67185-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 23-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Cytohesin 2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Cytohesin 2. This antibody recognizes human, mouse, rat antigen. The Cytohesin 2 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Cytohesin 2 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
9266 | |
Primary |
3A7A1 | |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Rat, Mouse | |
IgG2a |