missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 14 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22221-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specifically against KRT14.
Cytokeratins (CK) are intermediate filaments of epithelial cells, both in keratinizing tissue (ie. skin) and non-keratinizing cells (ie. mesothelial cells). At least 20 different CKs can be identified. Biochemically, most members of the CK family fall into one of two classes, type I (acidic polypeptides) and type II (basic polypeptides). Belonging to the type I subfamily of low molecular weight keratins and existing in combination with keratin 5, keratin 14 distinguishes stratified epithelial cells from simple epithelial cells and is useful in identification of squamous cell carcinomas. It is considered a prognostic marker in breast carcinomas. At least one member of the acidic family and one member of the basic family is expressed in all epithelial cells.Specifications
Cytokeratin 14 | |
Polyclonal | |
Unconjugated | |
KRT14 | |
CK 14, CK14, Cytokeratin 14, EBS3, EBS4, K14, keratin 14, KRT14, NFJ | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16664, 3861 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02533, Q61781 | |
Krt14 | |
Cytokeratin 14 Fusion Protein Ag17559 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |