missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 6 Mouse, Clone: KRT6/1702, enQuireBio™
Mouse Monoclonal Antibody
2035.00 SEK - 5730.00 SEK
Specifications
Antigen | Cytokeratin 6 |
---|---|
Clone | KRT6/1702 |
Concentration | 200μg/mL |
Applications | Immunohistochemistry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960318
|
EnquireBio
3853-MSM2-P1 |
100 μg |
5730.00 SEK
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950318
|
EnquireBio
3853-MSM2-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Cytokeratin 6 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
286887 | |
Recombinant human full-length keratin 6 protein | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
56kDa |
KRT6/1702 | |
Immunohistochemistry | |
Unconjugated | |
RUO | |
NP_775109 | |
KRT6A ;KRT6B ;KRT6C | |
IgG2a κ | |
Protein A/G Purified | |
Cytoplasmic | |
Tonsil or basal cell carcinoma |