missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 6A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16853-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody is against all three isoforms of keratin 6, but it does not bind other keratins.
Keratins 6 and 16 are expressed in keratinocytes, which are undergoing rapid turnover in the suprabasal region (also known as hyperproliferation-related keratins). Keratin 6 is found in hair follicles, suprabasal cells of a variety of internal stratified epithelia, in epidermis, in both normal and hyperproliferative situations. Epidermal injury results in activation of keratinocytes which express CK6 and CK16. CK6 is strongly expressed in about 75% of head and neck squamous cell carcinomas. Expression of CK6 is particularly associated with differentiation.Specifications
Cytokeratin 6A | |
Polyclonal | |
Unconjugated | |
KRT6A | |
CK 6A, CK 6D, CK6A, CK6C, CK6D, Cytokeratin 6A, Cytokeratin 6D, Hom s 5, K6A, K6C, K6D, keratin 6A, KRT conserve, KRT6, KRT6A, KRT6C, KRT6D, Type II keratin Kb6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16687, 3853 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P02538, P50446 | |
KRT6A | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |