missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 6B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17391-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. As many as six of this type II cytokeratin (KRT6) have been identified; the multiplicity of the genes is attributed to successive gene duplication events. The genes are expressed with family members KRT16 and/or KRT17 in the filiform papillae of the tongue, the stratified epithelial lining of oral mucosa and esophagus, the outer root sheath of hair follicles, and the glandular epithelia. Mutations in these genes have been associated with pachyonychia congenita. The type II cytokeratins are clustered in a region of chromosome 12q12-q13.Specifications
Cytokeratin 6B | |
Polyclonal | |
Unconjugated | |
KRT6B | |
CK 6B, CK6B, Cytokeratin 6B, K6B, keratin 6B, KRT6B, KRTL1, PC2, Type II keratin Kb10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16688, 3854 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04259, Q9Z331 | |
Krt6b | |
Cytokeratin 6B Fusion Protein Ag11288 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |