missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin, Acidic Rabbit, Clone: KRTL/1577R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio RBM21-1577-P1
Additional Details : Weight : 0.01000kg
Specifications
Cytokeratin, Acidic | |
Cytoplasmic | |
200μg/mL | |
10mM PBS with or without 0.05% BSA and 0.05% azide. | |
KRT77 | |
Recombinant human KRT77 protein fragment | |
56.5kDa (CK10); 50kDa (CK14); 50kDa (CK15); 48kDa (CK16); 40kDa (CK19) | |
Protein A/G Purified | |
RUO | |
Primary | |
374454 |
Immunohistochemistry | |
KRTL/1577R | |
Unconjugated | |
XP_011536590 | |
Rabbit | |
IgG kappa | |
Skin, Squamous cell carcinoma (SCC) | |
100 μg | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
Monoclonal |