Läs mer
CZC-8004, MedChemExpress
Produktkod. 16497127
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16497127 5 mg
16484408 10 mg
16443066 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 16497127 Leverantör Medchem Express Leverantörsnummer HY111138
MedChemExpress CZC-8004 is a pan-kinase inhibitor and binds a range of tyrosine kinases, including ABL kinase.
CZC-8004 is a pan-kinase inhibitor and binds a range of tyrosine kinases, including ABL kinase.
Specifikationer
Kemiskt namn eller material CZC-8004
Fysisk form Solid
CAS 916603-07-1
Färg White
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C17H16FN5
För användning med (applikation) Cancer-Kinase/protease
Synonym CZC-00008004
Löslighetsinformation DMSO : 77.5 mg/mL (250.53 mM; Need ultrasonic)
LEDER FC1=CN=C(NC2=CC=C(CN)C=C2)N=C1NC3=CC=CC=C3
Molekylvikt (g/mol) 309.34
Formel vikt 309.34
Procent renhet 99.61%