Läs mer
D-2’-Deoxyribofuranosyl-3-guanylurea(Alpha/Beta-Mixture), TRC
Produktkod. 30133642
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133642 50 mg
30133570 25 mg
30006099 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133642 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD25350050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 570410-72-9
Molekylformel C7H14N4O4
Molekylvikt (g/mol) 218.21
Synonym N-(Aminoiminomethyl)-N'-(2-deoxy-D-erythro-pentofuranosyl)urea;
IUPAC-namn N-(Aminoiminomethyl)-N'-(2-deoxy-D-erythro-pentofuranosyl)urea
LEDER O[C@H]1CC(NC(NC(N)=N)=O)O[C@@H]1CO
Specifikationer
Kemiskt namn eller material D-2'-Deoxyribofuranosyl-3-guanylurea
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C7H14N4O4/c8-6(9)11-7(14)10-5-1-3(13)4(2-12)15-5/h3-5,12-13H,1-2H2,(H5,8,9,10,11,14)/t3-,4+,5+/m0/s1
Formel vikt 218.1
Procent renhet >95%