Läs mer
D-(+)-2-Phosphoglyceric Acid Sodium Hydrate (~90%), TRC
Produktkod. 30009445
Change view
Kvantitet:
2.5 mg
10 mg
50 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30009445 2.5 mg 2.5mg
30006969 50 mg -
30005585 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009445 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3580002.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70195-25-4
Molekylformel C3 H5 O7 P . 2 Na . H2 O
Molekylvikt (g/mol) 248.04
IUPAC-namn disodium;[(1R)-1-carboxy-2-hydroxyethyl] phosphate;hydrate
LEDER O.[Na+].[Na+].OC[C@@H](OP(=O)([O-])[O-])C(=O)O
Specifikationer
Kemiskt namn eller material D-(+)-2-Phosphoglyceric Acid Sodium Hydrate
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C3H7O7P.2Na.H2O/c4-1-2(3(5)6)10-11(7,8)9;;;/h2,4H,1H2,(H,5,6)(H2,7,8,9);;;1H2/q;2*+1;/p-2/t2-;;;/m1.../s1
Formel vikt 247.9674
Procent renhet >95%