Läs mer
D-(-)-3-Phosphoglyceric Acid Disodium Salt, TRC
Produktkod. 30240773
Change view
Kvantitet:
25 mg
50 mg
200 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240773 200 mg
30240582 25 mg
30240787 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30240773 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP358203200MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 80731-10-8
Molekylformel C3H5O7PNa2
Molekylvikt (g/mol) 230.02
Synonym Propanoic acid, 2-hydroxy-3-(phosphonooxy)-, sodium salt (1:2), (2R)-, Propanoic acid, 2-hydroxy-3-(phosphonooxy)-, disodium salt, (R)- (9CI), Disodium D-3-phosphoglycerate, 3-Phospho-D-glyceric acid disodium salt, D-(-)-3-Phosphoglyceric acid disodium salt, (2R)-2-Hydroxy-3-(phosphonooxy)propanoic acid disodium salt
IUPAC-namn disodium;(2R)-2-hydroxy-3-[hydroxy(oxido)phosphoryl]oxypropanoate
LEDER [Na+].[Na+].O[C@H](COP(=O)(O)[O-])C(=O)[O-]
Specifikationer
Kemiskt namn eller material D-(-)-3-Phosphoglyceric Acid Disodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 200 mg
InChI formel InChI=1S/C3H7O7P.2Na/c4-2(3(5)6)1-10-11(7,8)9;;/h2,4H,1H2,(H,5,6)(H2,7,8,9);;/q;2*+1/p-2/t2-;;/m1../s1
Formel vikt 229.9568
Procent renhet >95%