Läs mer
D-Abequose, TRC
Produktkod. 30179337
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30179337 10 mg -
30179133 25 mg -
30179589 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179337 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA01020510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 56816-60-5
Molekylformel C6 H12 O4
Molekylvikt (g/mol) 148.16
Synonym Abequose (6CI), 3,6-Dideoxy-D-xylo-hexose;
IUPAC-namn (2R,4R,5R)-2,4,5-trihydroxyhexanal
LEDER C[C@@H](O)[C@H](O)C[C@@H](O)C=O
Specifikationer
Kemiskt namn eller material D-Abequose
Kvantitet 10 mg
InChI formel InChI=1S/C6H12O4/c1-4(8)6(10)2-5(9)3-7/h3-6,8-10H,2H2,1H3/t4-,5-,6-/m1/s1
Formel vikt 148.0736