missing translation for 'onlineSavingsMsg'
Learn More

D-Alaninamide hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-7055049
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
71810-97-4
C3H9ClN2O
124.568
MFCD00039093
FIAINKIUSZGVGX-HSHFZTNMSA-N
r-2-aminopropanamide hydrochloride, d-alaninamide hydrochloride, 2r-2-aminopropanamide hydrochloride, h-d-ala-nh2.hcl, d-alanine amide hydrochloride, propanamide, 2-amino-, monohydrochloride, 2r, souhmxhaa fjax@, pubchem12667, h-d-ala-nh2 hcl, d-ala-nh2 hcl
2775814
(2R)-2-aminopropanamide;hydrochloride
CC(C(=O)N)N.Cl
This item is not returnable. View return policy
71810-97-4
C3H9ClN2O
124.568
MFCD00039093
FIAINKIUSZGVGX-HSHFZTNMSA-N
r-2-aminopropanamide hydrochloride, d-alaninamide hydrochloride, 2r-2-aminopropanamide hydrochloride, h-d-ala-nh2.hcl, d-alanine amide hydrochloride, propanamide, 2-amino-, monohydrochloride, 2r, souhmxhaa fjax@, pubchem12667, h-d-ala-nh2 hcl, d-ala-nh2 hcl
2775814
(2R)-2-aminopropanamide;hydrochloride
CC(C(=O)N)N.Cl

D-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals.

Solubility
Soluble in 1% acetic acid and water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Color White
Solubility Information Soluble in 1% acetic acid and water.
Formula Weight 124.57
Percent Purity 98%
Quantity 1 g
Physical Form Powder
Chemical Name or Material D-Alaninamide hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'