missing translation for 'onlineSavingsMsg'
Läs mer

D-Alanine-3,3,3-d3, TRC

Produktkod. 30179526
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179526 50 mg
30179163 100 mg
30179080 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Reglering: Begränsningar kan gälla för köp av denna produkt och vi kan behöva begära ytterligare dokumentation från dig.
Produktkod. 30179526 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA48099850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 177614-69-6
Molekylformel C3 D3 H4 N O2
Molekylvikt (g/mol) 92.1117
Synonym D-Alanine-3,3,3-d3 (9CI, ACI)
IUPAC-namn (2R)-2-amino-3,3,3-trideuteriopropanoic acid
LEDER [2H]C([2H])([2H])[C@@H](N)C(=O)O

Specifications

Kemiskt namn eller material D-Alanine-3,3,3-d3
Kvantitet 50 mg
InChI formel InChI=1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m1/s1/i1D3
Formel vikt 92.0665
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.