Läs mer
D-Allocystathionine, TRC
Produktkod. 30313020
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30313020 10 mg
1 options
Produktkod. 30313020 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA54612510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material D-Allocystathionine
CAS 2998-83-6
Molekylformel C7 H14 N2 O4 S
Kvantitet 10 mg
Synonym D-Homocysteine, S-(2-amino-2-carboxyethyl)-, (R)- (9CI), Allocystathionine, D- (8CI), (-)-Allocystathionine, D-Allocystathionine, S-[(2R)-2-Amino-2-carboxyethyl]-D-homocysteine, (2R)-2-Amino-4-[[(2R)-2-amino-2-carboxyethyl]thio]butanoic acid
InChI formel InChI=1S/C7H14N2O4S/c8-4(6(10)11)1-2-14-3-5(9)7(12)13/h4-5H,1-3,8-9H2,(H,10,11)(H,12,13)/t4-,5-/m1/s1
LEDER N[C@H](CCSC[C@@H](N)C(=O)O)C(=O)O
IUPAC-namn (2R)-2-amino-4-[(2S)-2-amino-2-carboxyethyl]sulfanylbutanoic acid
Molekylvikt (g/mol) 222.26
Formel vikt 222.0674