Läs mer
D-alpha-Tocopherol Quinone, TRC
Produktkod. 30455951
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455951 1 g
30453313 100 mg
30454917 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30455951 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT5262101G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7559-04-8
Molekylformel C29H50O3
Molekylvikt (g/mol) 446.71
IUPAC-namn 2-[(3R,7R,11R)-3-hydroxy-3,7,11,15-tetramethylhexadecyl]-3,5,6-trimethylcyclohexa-2,5-diene-1,4-dione
LEDER O=C1C(C)=C(C)C(C(CCC(O)(C)CCCC(C)CCCC(C)CCCC(C)C)=C1C)=O
Specifikationer
Kemiskt namn eller material D-alpha-Tocopherol Quinone
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
Synonym [3R-(3R*,7R*,11R*)]-2-(3-Hydroxy-3,7,11,15-tetramethylhexadecyl)-3,5,6-trimethyl-2,5-cyclohexadiene-1,4-dione, 2-(3-Hydroxy-3,7,11,15-tetramethylhexadecyl)-3,5,6-trimethyl-p-benzoquinone, 2-[(3R,7R,11R)-3-Hydroxy-3,7,11,15-tetramethylhexadecyl]-3,5,6-trimethyl-2,5-cyclohexadiene-1,4-dione, 2-((3R,7R,11R)-3-Hydroxy-3,7,11,15-tetramethylhexadecyl)-3,5,6-trimethylcyclohexa-2,5-diene-1,4-dione, Metarene, Tocopherol Quinone, Tocopherolquine, Tocopheryl Quinone, Vitamin E Quinone, α-Tocopherol Quinone, α-Tocopherylquinone, α-Tocoquinone
InChI formel InChI=1S/C29H50O3/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8,32)19-17-26-25(7)27(30)23(5)24(6)28(26)31/h20-22,32H,9-19H2,1-8H3/t21-,22-,29-/m1/s1
Formel vikt 446.376
Procent renhet >95%