missing translation for 'onlineSavingsMsg'
Learn More
Learn More
d-alpha-Tocopheryl Retinoate, TRC
Specifications
Specifications
| Kemiskt namn eller material | d-alpha-Tocopheryl Retinoate |
| CAS | 40516-49-2 |
| Molekylformel | C49H76O3 |
| Kvantitet | 500 mg |
| Synonym | Retinoic Acid (2R)-3,4-Dihydro-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-benzopyran-6-yl Ester, [2R-[2R*(4R*,8R*)]]-Retinoic Acid 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-1-benzopyran-6-yl Ester, d-α-Tocopherol Retinoate, α-Tocopheryl Retinoate |
| InChI formel | InChI=1S/C49H76O3/c1-34(2)19-14-20-35(3)21-15-22-36(4)25-17-31-49(13)32-29-43-42(10)46(40(8)41(9)47(43)52-49)51-45(50)33-38(6)24-16-23-37(5)27-28-44-39(7)26-18-30-48(44,11)12/h16,23-24,27-28,33-36H,14-15,17-22,25-26,29-32H2,1-13H3/b24-16+,28-27+,37-23+,38-33+/t35-,36-,49-/m1/s1 |
| LEDER | O=C(OC1=C(C)C(CC[C@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)O2)=C2C(C)=C1C)/C=C(C)/C=C/C=C(C)/C=C/C3=C(C)CCCC3(C)C |
| IUPAC-namn | (R)-2,5,7,8-tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-yl (2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoate |
| Molekylvikt (g/mol) | 713.13 |
| Formel vikt | 712.58 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?