missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Arabinose, ≥99%, MP Biomedicals™

Catalog Number p-4403817
Click to view available options
Quantity:
25 g
100 g
1 kg
This item is not returnable. View return policy
28697-53-2
C5H10O5
150.13
MFCD00064361,MFCD20731172,MFCD00006609,MFCD00066648,MFCD09864633
PYMYPHUHKUWMLA-DEOSMSJNSA-N
alpha-d-arabinopyranose, 2s,3s,4r,5r-oxane-2,3,4,5-tetrol, d--arabinose, alpha-arabinose, a-d-arabinopyranose, alpha-d-arabinopyranose 9ci, 2s,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol
71313478
(2S,3R)-2,3,4,5-tetrahydroxypentanal
OCC(O)[C@@H](O)[C@H](O)C=O
This item is not returnable. View return policy
28697-53-2
C5H10O5
150.13
MFCD00064361,MFCD20731172,MFCD00006609,MFCD00066648,MFCD09864633
PYMYPHUHKUWMLA-DEOSMSJNSA-N
alpha-d-arabinopyranose, 2s,3s,4r,5r-oxane-2,3,4,5-tetrol, d--arabinose, alpha-arabinose, a-d-arabinopyranose, alpha-d-arabinopyranose 9ci, 2s,3s,4r,5r-tetrahydro-2h-pyran-2,3,4,5-tetraol
71313478
(2S,3R)-2,3,4,5-tetrahydroxypentanal
OCC(O)[C@@H](O)[C@H](O)C=O

D-(—)-Arabinose is a substrate used to identify, differentiate and characterize arabinose isomerase(s) that have commercial value in the production of tagatose, a low calorie sweetener

Color White
Assay Percent Range ≥99%
Quantity 25 g
Formula Weight 150.13
Physical Form Powder
Chemical Name or Material D-(-)-Arabinose
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'