Läs mer
D-Aspartic Acid 1-(1,1-Dimethylethyl) Ester, TRC
Produktkod. 30178931
Change view
Kvantitet:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30178931 1 g
30179578 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30178931 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA7887951G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 148823-36-3
Molekylformel C8H15NO4
Molekylvikt (g/mol) 189.21
Synonym D-Aspartic Acid 1-tert-Butyl Ester
LEDER CC(C)(C)OC(=O)[C@@H](N)CC(=O)O
Specifikationer
Kemiskt namn eller material D-Aspartic Acid 1-(1,1-Dimethylethyl) Ester
Kvantitet 1 g