missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Biotin, MP Biomedicals
Click to view available options
Quantity:
100 mg
500 mg
Description
- D-Biotin may be used to elute proteins from avidin/streptavidin resins
- It is widely used for dietary supplements and fortified foods
- It is also used for tablets and hard-shell capsule preparation due to its pharmaceutical properties
Specifications
Melting Point | 223°C to 233°C |
Color | White |
Boiling Point | 573.6°C (760 mmHg(literature)) |
Linear Formula | C10H16N2O3S |
Quantity | 100 mg |
Solubility Information | Soluble in water (22 mg/100 mL), ethanol (80 mg/100 mL), more soluble in hot water and in dilute alkalies; insoluble in other common organic solvents. Soluble in 2 M Ammonium hydroxide (50mg/mL - clear, colorless solution), dimethylformamide (1.7mg/mL). |
Formula Weight | 244.3 |
Percent Purity | ≥97.5% |
Physical Form | Crystalline Powder |
Chemical Name or Material | D-BIOTIN |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'