Läs mer
D-Citronellol, TRC
Produktkod. 17913385
Change view
Kvantitet:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17913385 1 g
17924905 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17913385 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC5250351G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1117-61-9
Molekylformel C10 H20 O
Molekylvikt (g/mol) 156.27
Synonym (R)-(+)-β-Citronellol, (3R)-3,7-dimethyl-6-octen-1-ol
IUPAC-namn (3R)-3,7-dimethyloct-6-en-1-ol
LEDER C[C@@H](CCO)CCC=C(C)C
Specifikationer
Kemiskt namn eller material D-Citronellol
Kvantitet 1 g
InChI formel InChI=1S/C10H20O/c1-9(2)5-4-6-10(3)7-8-11/h5,10-11H,4,6-8H2,1-3H3/t10-/m1/s1
Formel vikt 156.1514