Läs mer
D-Cysteine Methyl Ester Hydrochloride, TRC
Produktkod. 30367147
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30367147 250 mg
1 options
Produktkod. 30367147 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC995300250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material D-Cysteine Methyl Ester Hydrochloride
CAS 70361-61-4
Molekylformel C4H10ClNO2S
Kvantitet 250 mg
Synonym (S)-Cysteine Methyl Ester Hydrochloride, Methyl D-Cysteinate Hydrochloride
InChI formel InChI=1S/C4H9NO2S.ClH/c1-7-4(6)3(5)2-8;/h3,8H,2,5H2,1H3;1H/t3-;/m1./s1
LEDER N[C@H](CS)C(OC)=O.Cl
IUPAC-namn methyl D-cysteinate hydrochloride
Molekylvikt (g/mol) 171.65
Formel vikt 171.01