missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Dextrose, Anhydrous, Thermo Scientific Chemicals

Catalog Number p-7082785
Click to view available options
Quantity:
5 lb.
25 lb.
100 lb.
Packaging:
Plastic bottle
This item is not returnable. View return policy
50-99-7
C6H12O6
180.156
MFCD00063774
GZCGUPFRVQAUEE-SLPGGIOYSA-N
2 r,3 s,4 r,5 r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
107526
CHEBI:42758
(2 R,3 S,4 R,5 R)-2,3,4,5,6-pentahydroxyhexanal
C(C(C(C(C(C=O)O)O)O)O)O
This item is not returnable. View return policy
50-99-7
C6H12O6
180.156
MFCD00063774
GZCGUPFRVQAUEE-SLPGGIOYSA-N
2 r,3 s,4 r,5 r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
107526
CHEBI:42758
(2 R,3 S,4 R,5 R)-2,3,4,5,6-pentahydroxyhexanal
C(C(C(C(C(C=O)O)O)O)O)O
Melting Point 147°C to 153°C
Packaging Plastic bottle
Linear Formula CH2OH(CHOH)4CHO
Quantity 5 lb.
Beilstein 1281608
Formula Weight 180.16
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Dextrose

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'