missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
D-Dimer Antibody (28), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-05045DL594
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
D-Dimer Monoclonal antibody specifically detects D-Dimer in Human, Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)
Specifikationer
| D-Dimer | |
| Monoclonal | |
| DyLight 594 | |
| D Dimer | |
| Purified human D-dimer | |
| 0.1 mL | |
| Extracellular Matrix | |
| Human, Canine | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
| 28 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction