Läs mer
D-DOPA, TRC
Produktkod. 17959744
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17959744 250 mg
17999354 500 mg
17999454 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17959744 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD533750250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5796-17-8
Molekylformel C9H11NO4
Molekylvikt (g/mol) 197.19
Synonym D-Tyrosine, 3-hydroxy-, Alanine, 3-(3,4-dihydroxyphenyl)-, D- (8CI), 3-Hydroxy-D-tyrosine, (+)-3-(3,4-Dihydroxyphenyl)alanine, (+)-Dopa, 3,4-Dihydroxy-D-phenylalanine, 3-(3,4-Dihydroxyphenyl)-D-alanine, D-3,4-Dihydroxyphenylalanine, D-3-(3,4-Dihydroxyphenyl)alanine, D-3-Hydroxytyrosine, D-Dopa, β-(3,4-Dihydroxy)-D-phenylalanine, (2R)-2-Amino-3-(3,4-dihydroxyphenyl)propanoic Acid
LEDER N[C@H](Cc1ccc(O)c(O)c1)C(=O)O
Specifikationer
Kemiskt namn eller material D-DOPA
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12H,3,10H2,(H,13,14)/t6-/m1/s1
Kvantitet 250 mg
Formel vikt 197.0688
Procent renhet >95%