Läs mer
D-erythro-Chloramphenicol, TRC
Produktkod. 17929629
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17929629 10 mg
18073310 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17929629 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC32500510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7387-98-6
Molekylformel C11 H12 Cl2 N2 O5
Molekylvikt (g/mol) 323.13
Synonym [S-(R*,S*)]-2,2-Dichloro-N-[2-hydroxy-1-(hydroxymethyl)-2-(4-nitrophenyl)ethyl]-acetamide, D-erythro-2,2-Dichloro-N-[β-hydroxy-α-(hydroxymethyl)-p-nitrophenethyl]-acetamide, D-erythro-(1S,2R)-1-p-Nitrophenyl-2-dichloroacetamido-1,3-propanediol
IUPAC-namn 2,2-dichloro-N-[(1R,2S)-2-hydroxy-1-(hydroxymethyl)-2-(4-nitrophenyl)ethyl]acetamide
LEDER OC[C@@H](NC(=O)C(Cl)Cl)[C@@H](O)c1ccc(cc1)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material D-erythro-Chloramphenicol
Kvantitet 10 mg
InChI formel InChI=1S/C11H12Cl2N2O5/c12-10(13)11(18)14-8(5-16)9(17)6-1-3-7(4-2-6)15(19)20/h1-4,8-10,16-17H,5H2,(H,14,18)/t8-,9+/m1/s1
Formel vikt 322.0123