Läs mer
D-erythro-Sphingosylphosphorylcholine (Synthetic), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | D-Erythro-Sphingosylphosphorylcholine |
| CAS | 1670-26-4 |
| Molekylformel | C23 H49 N2 O5 P |
| Kvantitet | 1 mg |
| Synonym | Ethanaminium','2-[[[[(2S,3R,4E)-2-amino-3-hydroxy-4-octadecen-1-yl]oxy]hydroxyphosphinyl]oxy]-N,N,N-trimethyl-','inner salt, Choline','hydroxide','2-amino-3-hydroxy-4-octadecenyl hydrogen phosphate','inner salt','(E)-D-erythro- (8CI), Ethanaminium','2-[[[(2-amino-3-hydroxy-4-octadecenyl)oxy]hydroxyphosphinyl]oxy]-N,N,N-trimethyl-','inner salt','[R-[R*,S*-(E)]]-, Ethanaminium','2-[[[[(2S,3R,4E)-2-amino-3-hydroxy-4-octadecenyl]oxy]hydroxyphosphinyl]oxy]-N,N,N-trimethyl-','inner salt (9CI), C18-Sphingosine phosphocholine, Lysosphingomyelin, Sphingenyl-1-phosphorylcholine, Sphingosylphosphorylcholine, erythro-Sphingosylphosphorylcholine |
| InChI formel | InChI=1S/C23H49N2O5P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(26)22(24)21-30-31(27,28)29-20-19-25(2,3)4/h17-18,22-23,26H,5-16,19-21,24H2,1-4H3/b18-17+/t22-,23+/m0/s1 |
| LEDER | CCCCCCCCCCCCC\C=C\[C@@H](O)[C@@H](N)COP(=O)([O-])OCC[N+](C)(C)C |
| IUPAC-namn | [(E,2S,3R)-2-amino-3-hydroxyoctadec-4-enyl] 2-(trimethylazaniumyl)ethyl phosphate |
| Molekylvikt (g/mol) | 464.62 |
| Formel vikt | 464.3379 |