Läs mer
D-Erythrose (w/v = 10 mg/ml), TRC
Produktkod. 17997534
Change view
Kvantitet:
1 mL
2.5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17997534 1 mL
17987584 2.5 mL
17939264 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17997534 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE6501301ML
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 583-50-6
Molekylformel C4 H8 O4
Molekylvikt (g/mol) 120.1
Synonym Butanal, 2,3,4-trihydroxy-, (2R,3R)-, Butanal, 2,3,4-trihydroxy-, [R-(R*,R*)]-, Erythrose, D- (8CI), (2R,3R)-2,3,4-Trihydroxybutanal, D-(-)-Erythrose, D-Erythrose
IUPAC-namn (2R,3R)-2,3,4-trihydroxybutanal
LEDER OC[C@@H](O)[C@@H](O)C=O
Specifikationer
Kemiskt namn eller material D-(-)-Erythrose
Kvantitet 1 mL
InChI formel InChI=1S/C4H8O4/c5-1-3(7)4(8)2-6/h1,3-4,6-8H,2H2/t3-,4+/m0/s1
Formel vikt 120.0423