Läs mer
D-Erythrulose (1M in Water), 90%, TRC
Produktkod. 30179663
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179663 5 mg
30179631 50 mg
30179506 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179663 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE6501465MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 496-55-9
Molekylformel C4 H8 O4
Molekylvikt (g/mol) 120.1
Synonym 2-Butanone, 1,3,4-trihydroxy-, (3R)-, 2-Butanone, 1,3,4-trihydroxy-, (R)-, D-glycero-Tetrulose (7CI,8CI), (3R)-1,3,4-Trihydroxy-2-butanone, D-Erythrulose, D-Threulose, D-glycero-2-Tetrulose
IUPAC-namn (3R)-1,3,4-trihydroxybutan-2-one
LEDER OC[C@@H](O)C(=O)CO
Specifikationer
Kemiskt namn eller material D-Erythrulose
Kvantitet 5 mg
InChI formel InChI=1S/C4H8O4/c5-1-3(7)4(8)2-6/h3,5-7H,1-2H2/t3-/m1/s1
Formel vikt 120.0423
Koncentration eller sammansättning (efter analyt eller komponenter) D-Erythrulose 1M