Läs mer
D-Galactono-1,4-lactone, TRC
Produktkod. 30401324
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30401324 100 mg
1 options
Produktkod. 30401324 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG155400100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material D-Galactono-1,4-lactone
CAS 2782-07-2
Molekylformel C6 H10 O6
Kvantitet 100 mg
Synonym D-Galactonic acid, γ-lactone, Galactonic acid, γ-lactone, D- (8CI), 1,4-D-Galactonolactone, D-Galactonic acid 1,4-lactone, D-Galactono-1,4-lactone, D-Galactono-γ-lactone, NSC 102762, γ-D-Galactonolactone
InChI formel InChI=1S/C6H10O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2-5,7-10H,1H2/t2-,3-,4-,5+/m1/s1
LEDER OC[C@@H](O)[C@@H]1OC(=O)[C@H](O)[C@H]1O
IUPAC-namn (3R,4R,5S)-5-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
Molekylvikt (g/mol) 178.14
Formel vikt 178.0477