Läs mer
D-Galactose-13C6, TRC
Produktkod. 30005028
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005028 50 mg
30012668 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30005028 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG15525150MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 74134-89-7
Molekylformel 13C6H12O6
Molekylvikt (g/mol) 186.11
Synonym (+)-Galactose-13C6, D-(+)-Galactose-13C6, Galactose-13C6
LEDER O[13C]([H])([H])[13C@@]1([H])O[13C](O)([H])[13C@@](O)([H])[13C@](O)([H])[13C@@]1([H])O
Specifikationer
Kemiskt namn eller material D-Galactose-13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vikt 186.0835
Procent renhet >95%