Läs mer
D-Glucoheptono-1,4-lactone, TRC
Produktkod. 30404216
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30404216 25 mg
1 options
Produktkod. 30404216 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG41650025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material D-Glucoheptono-1,4-lactone
CAS 60046-25-5
Molekylformel C7 H12 O7
Kvantitet 25 mg
Synonym (2ξ)-D-Glucoheptonic Acid γ-Lactone, Glucoheptonic Acid γ-Lactone, D-Glucoheptonolactone
InChI formel InChI=1S/C7H12O7/c8-1-2(9)3(10)6-4(11)5(12)7(13)14-6/h2-6,8-12H,1H2/t2-,3-,4+,5-,6+/m1/s1
LEDER OC[C@@H](O)[C@@H](O)[C@@H]1OC(=O)[C@H](O)[C@@H]1O
IUPAC-namn (3R,4S,5S)-3,4-dihydroxy-5-[(1R,2R)-1,2,3-trihydroxypropyl]oxolan-2-one
Molekylvikt (g/mol) 208.17
Formel vikt 208.0583