Learn More
Melford D-Gluconic Acid
Product Code. 30018295
Change view
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018295 500 g 500g
30018294 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30018295 Supplier Melford Supplier No. G37020500.0
 Restricted Item
Beräknad leverans: 04-12-2026
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Molecular Formula: C6H11O7K, MW 234.2
Specifications
Chemical Name or Material D-Gluconic Acid
CAS 299-27-4
Molecular Formula C6H11KO7
Quantity 500 g