Läs mer
D-Glucosamine 6-Phosphate Hydrate, TRC
Produktkod. 30240986
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240986 10 mg
30240703 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30240986 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG51505010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3616-42-0
Molekylformel C6H16NO9P
Molekylvikt (g/mol) 277.17
Synonym 2-Amino-2-deoxy-D-glucose 6-(Dihydrogen Phosphate) Hydrate, Glucosamine 6-Phosphate Hydrate, 2-Amino-2-deoxy-D-glucose 6-Phosphate Hydrate, 2-Amino-2-deoxyglucose 6-Phosphate Hydrate, 2-Amino-D-glucose-6-phosphate Hydrate, D-Glucosamine 6-Phosphate Hydrate;
IUPAC-namn [(2R,3S,4R,5R)-5-amino-3,4,6-trihydroxyoxan-2-yl]methyl dihydrogen phosphate
LEDER N[C@H]1C(O)O[C@H](COP(=O)(O)O)[C@@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material D-Glucosamine 6-Phosphate Hydrate
Kvantitet 10 mg
InChI formel InChI=1S/C6H14NO8P/c7-3-5(9)4(8)2(15-6(3)10)1-14-16(11,12)13/h2-6,8-10H,1,7H2,(H2,11,12,13)/t2-,3-,4-,5-,6?/m1/s1
Formel vikt 259.0457