Läs mer
D-Glucose-1-13C, TRC
Produktkod. 30179354
Change view
Kvantitet:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179354 500 mg
30179622 250 mg
30179180 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179354 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG595007500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 40762-22-9
Molekylformel 13C C5 H12 O6
Molekylvikt (g/mol) 181.15
Synonym D-Glucose-1-13C, 1-13C-Glucose, D-[1-13C]Glucose, Glucose-1-13C, [1-13C]-D-Glucose, [1-13C]D-Glucose
IUPAC-namn (3R,4S,5S,6R)-6-(hydroxymethyl)(2^{13}C)oxane-2,3,4,5-tetrol
LEDER OC[C@H]1O[13CH](O)[C@H](O)[C@@H](O)[C@@H]1O
Specifikationer
Kemiskt namn eller material D-Glucose 1-13C
Kvantitet 500 mg
InChI formel InChI=1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i6+1
Formel vikt 181.0667