Läs mer
D-Glucose-2-13C, TRC
Produktkod. 30178920
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30178920 25 mg
30179643 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30178920 Leverantör Toronto Research Chemicals Leverantörsnummer TRCG59500825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 105931-74-6
Molekylformel C5 13C H12 O6
Molekylvikt (g/mol) 181.15
Synonym 2-13C-Glucose, D-[2-13C]Glucose, Glucose-2-13C, [2-13C]-D-Glucose, [2-13C]D-Glucose
LEDER OC[C@H]1O[C@H](O)[13C@H](O)[C@@H](O)[C@@H]1O ∣a:2,6,8,10,&1:4∣
Specifikationer
Kemiskt namn eller material D-Glucopyranose-2-13C
Kvantitet 25 mg
InChI formel InChI=1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i5+1
Formel vikt 181.0667