Läs mer
Thermo Scientific Chemicals D-Glucose dehydrogenase
Beskrivning
Diagnostics and cofactor regeneration of NAD(P)H in reductive processes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifikationer
Specifikationer
| Kemiskt namn eller material | D-Glucose dehydrogenase |
| CAS | 9028-53-9 |
| Färg | Yellow |
| MDL-nummer | MFCD00131181 |
| Kvantitet | 1 kU |
| Fysisk form | Powder |