Läs mer
(D-Glycero-Alpha-D-manno-heptopyranosyl)-dihydogenphosphate, Cyclohexylammonium Salt, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | (D-Glycero-a-D-manno-heptopyranosyl)-dihydogenphosphate, Cyc |
| CAS | 359435-45-3 |
| Molekylformel | C7 H15 O10 P . C6 H13 N |
| Kvantitet | 10 mg |
| Synonym | D-glycero-α-D-manno-Heptopyranose, 1-(dihydrogen phosphate), compd. with cyclohexanamine (1:1) (9CI), D-glycero-α-D-manno-Heptopyranose 1-phosphate cyclohexylammonium salt |
| InChI formel | InChI=1S/C7H15O10P.C6H13N/c8-1-2(9)6-4(11)3(10)5(12)7(16-6)17-18(13,14)15;7-6-4-2-1-3-5-6/h2-12H,1H2,(H2,13,14,15);6H,1-5,7H2/t2-,3+,4+,5+,6-,7-;/m1./s1 |
| LEDER | NC1CCCCC1.OC[C@@H](O)[C@H]2O[C@H](OP(=O)(O)O)[C@@H](O)[C@@H](O)[C@@H]2O |
| IUPAC-namn | cyclohexanamine;[(2R,3S,4S,5S,6R)-6-[(1R)-1,2-dihydroxyethyl]-3,4,5-trihydroxyoxan-2-yl] dihydrogen phosphate |
| Molekylvikt (g/mol) | 389.34 |
| Formel vikt | 389.1451 |