Läs mer
D-Histidine Hydrochloride Hydrate, TRC
Produktkod. 30179568
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30179568 500 mg
1 options
Produktkod. 30179568 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH456033500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material D-Histidine Hydrochloride Hydrate
CAS 328526-86-9
Molekylformel C6H9N3O2 . HCl . xH 2O
InChI formel InChI=1S/C6H9N3O2.ClH/c7-5(6(10)11)1-4-2-8-3-9-4;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H/t5-;/m1./s1
LEDER O=C([C@@H](CC1=CN=CN1)N)O.Cl.O
IUPAC-namn D-histidine hydrochloride hydrate
Molekylvikt (g/mol) 209.63