Läs mer
D-Histidine Methyl Ester, TRC
Produktkod. 30407950
Change view
Kvantitet:
2.5 g
5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30407950 2.5 g 2.5g
30403839 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30407950 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH4560302.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material D-Histidine Methyl Ester
CAS 17720-12-6
Molekylformel C7H11N3O2
Kvantitet 2.5 g
Synonym R-Histidine Methyl Ester
InChI formel InChI=1S/C7H11N3O2/c1-12-7(11)6(8)2-5-3-9-4-10-5/h3-4,6H,2,8H2,1H3,(H,9,10)/t6-/m1/s1
LEDER COC(=O)[C@H](N)Cc1cnc[nH]1
IUPAC-namn methyl (2R)-2-amino-3-(1H-imidazol-5-yl)propanoate
Molekylvikt (g/mol) 169.18
Formel vikt 169.0851