Läs mer
D,L-4'-Methyl-Alpha-(1-isopropylaminomethyl) Benzyl Alcohol, Hydrochloride, TRC
Produktkod. 30179181
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179181 10 g
30179179 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179181 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM31455010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 17267-27-5
Molekylformel C12 H19 N O . Cl H
Molekylvikt (g/mol) 229.75
IUPAC-namn 1-(4-methylphenyl)-2-(propan-2-ylamino)ethanol;hydrochloride
LEDER Cl.CC(C)NCC(O)c1ccc(C)cc1
Specifikationer
Kemiskt namn eller material D,L-4'-Methyl-a-(1-isopropylaminomethyl) Benzyl Alcohol, Hyd
Kvantitet 10 g
InChI formel InChI=1S/C12H19NO.ClH/c1-9(2)13-8-12(14)11-6-4-10(3)5-7-11;/h4-7,9,12-14H,8H2,1-3H3;1H
Formel vikt 229.1233