Läs mer
D,L-Cystathionine, TRC
Produktkod. 30137522
Change view
Kvantitet:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137522 100 mg
30134293 250 mg
30012648 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137522 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC989495100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 535-34-2
Molekylformel C7 H14 N2 O4 S
Molekylvikt (g/mol) 222.26
Synonym S-(2-Amino-2-carboxyethyl)-homocysteine, DL-Allocystathionine
IUPAC-namn 2-amino-4-(2-amino-2-carboxy-ethyl)sulfanyl-butanoic acid
LEDER NC(CCSCC(N)C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material DL-Cystathionine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C7H14N2O4S/c8-4(6(10)11)1-2-14-3-5(9)7(12)13/h4-5H,1-3,8-9H2,(H,10,11)(H,12,13)
Kvantitet 100 mg
Formel vikt 222.0674
Procent renhet >95%