Läs mer
D,L-Mevalonic Acid Dicyclohexylammonium Salt, TRC
Produktkod. 30472495
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30472495 50 mg
30480033 100 mg
30472496 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30472495 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33902050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1215802-31-5
Molekylformel C18H35NO4
Molekylvikt (g/mol) 329.47
Synonym 3,5-Dihydroxy-3-(methyl) Pentanoic Acid Dicyclohexylammonium Salt, Hiochic Acid Dicyclohexylammonium Salt, 3,5-Dihydroxy-3-(methyl)valeric Acid Dicyclohexylammonium Salt, MK 91, MVA, MVS, (RS)-Mevalonic Acid Dicyclohexylammonium Salt
IUPAC-namn N-cyclohexylcyclohexanamine;3,5-dihydroxy-3-methylpentanoic acid
LEDER CC(O)(CCO)CC(=O)O.C1CCC(CC1)NC2CCCCC2
Specifikationer
Kemiskt namn eller material D,L-Mevalonic Acid Dicyclohexylammonium Salt
Kvantitet 50 mg
InChI formel InChI=1S/C12H23N.C6H12O4/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12;1-6(10,2-3-7)4-5(8)9/h11-13H,1-10H2;7,10H,2-4H2,1H3,(H,8,9)
Formel vikt 329.2566