Läs mer
D,L-N,N-Didesmethyl-N-(4-methoxyphenethyl) Venlafaxine, TRC
Produktkod. 30133974
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133974 100 mg
17993865 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133974 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD441150100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1329795-88-1
Molekylformel C24 H33 N O3
Molekylvikt (g/mol) 383.52
Synonym 1-[(1RS)-1-(4-Methoxyphenyl)-2-[[2-(4-methoxyphenyl)ethyl]amino]ethyl]cyclohexanol, Venlafaxine Hydrochloride Imp. H (EP), Cyclohexanol, 1-[1-(4-methoxyphenyl)-2-[[2-(4-methoxyphenyl)ethyl]amino]ethyl]- (ACI), 1-[1-(4-Methoxyphenyl)-2-[[2-(4-methoxyphenyl)ethyl]amino]ethyl]cyclohexanol (ACI), 1-[1-(4-Methoxyphenyl)-2-[2-(4-methoxyphenyl)ethylamino]ethyl]cyclohexan-1-ol
IUPAC-namn 1-[1-(4-methoxyphenyl)-2-[2-(4-methoxyphenyl)ethylamino]ethyl]cyclohexan-1-ol
LEDER COc1ccc(CCNCC(c2ccc(OC)cc2)C3(O)CCCCC3)cc1
Specifikationer
Kemiskt namn eller material 1-[(1RS)-1-(4-Methoxyphenyl)-2-[(2-(4-methoxyphenyl)ethyl)amino]ethyl]cyclohexanol
Kvantitet 100 mg
InChI formel InChI=1S/C24H33NO3/c1-27-21-10-6-19(7-11-21)14-17-25-18-23(24(26)15-4-3-5-16-24)20-8-12-22(28-2)13-9-20/h6-13,23,25-26H,3-5,14-18H2,1-2H3
Formel vikt 383.246