Learn More
D,L-O-Desmethyl Venlafaxine-d6, TRC
Product Code. 30361138
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity Unit Size
30361138 10 mg 10mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30361138 Supplier Toronto Research Chemicals Supplier No. TRCD29650210MG
 Restricted Item
Beräknad leverans: 10-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material D,L-O-Desmethyl Venlafaxine-d6
CAS 1020719-35-0
Purity Grade Notes HPLC
Molecular Formula C16H19D6NO2
Quantity 10 mg
Synonym 4-[2-(Dimethylamino)-1-(1-hydroxycyclo-hexyl)ethyl]phenol-d6
InChI Formula InChI=1S/C16H25NO2/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16/h6-9,15,18-19H,3-5,10-12H2,1-2H3/i3D2,4D2,5D2
SMILES OC1(CC([2H])([2H])C([2H])([2H])C([2H])([2H])C1)C(CN(C)C)C2=CC=C(O)C=C2
IUPAC Name 3,3,4,4,5,5-hexadeuterio-1-[2-(dimethylamino)-1-(4-hydroxyphenyl)ethyl]cyclohexan-1-ol
Molecular Weight (g/mol) 269.41
Formula Weight 269.2262
Percent Purity >95