Läs mer
D,L-Sulforaphane N-Acetyl-L-cysteine, TRC
Produktkod. 30008840
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008840 10 mg
30006543 1 mg
30003782 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008840 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS69912010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 334829-66-2
Molekylformel C11 H20 N2 O4 S3
Molekylvikt (g/mol) 340.48
IUPAC-namn (2R)-2-acetamido-3-(4-methylsulfinylbutylcarbamothioylsulfanyl)propanoic acid
LEDER CC(=O)N[C@@H](CSC(=S)NCCCCS(=O)C)C(=O)O
Specifikationer
Kemiskt namn eller material DL-Sulforaphane-N-acetyl-L-cysteine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C11H20N2O4S3/c1-8(14)13-9(10(15)16)7-19-11(18)12-5-3-4-6-20(2)17/h9H,3-7H2,1-2H3,(H,12,18)(H,13,14)(H,15,16)/t9-,20?/m0/s1
Kvantitet 10 mg
Formel vikt 340.0585
Procent renhet >95%