Läs mer
D,L-Sulforaphane, TRC
Produktkod. 17969344
Change view
Kvantitet:
25 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17969344 1 g
17938304 25 mg
17959724 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17969344 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS6991151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4478-93-7
Molekylformel C6 H11 N O S2
Molekylvikt (g/mol) 177.29
Synonym Isothiocyanic acid, 4-(methylsulfinyl)butyl ester (8CI), Sulforaphan (6CI), 1-Isothiocyanato-4-(methylsulfinyl)butane, 4-Methylsulfinylbutyl isothiocyanate, BroccoPhane, Detoxophane, Sulforaphane, Butane, 1-isothiocyanato-4-(methylsulfinyl)-
IUPAC-namn 1-isothiocyanato-4-methylsulfinylbutane
LEDER CS(=O)CCCCN=C=S
Specifikationer
Kemiskt namn eller material DL-Sulforaphane
Kvantitet 1 g
InChI formel InChI=1S/C6H11NOS2/c1-10(8)5-3-2-4-7-6-9/h2-5H2,1H3
Formel vikt 177.0282