Läs mer
(D,L)-threo-alpha-Phenyl-2-piperidineacetamide, TRC
Produktkod. 17982845
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17982845 25 mg
17952645 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17982845 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP33597625MG
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 50288-62-5
Molekylformel C13 H18 N2 O
Molekylvikt (g/mol) 218.29
Synonym 2-Piperidineacetamide, α-phenyl-, (αR,2R)-rel-, 2-Piperidineacetamide, α-phenyl-, (R*,R*)-, (2RS)-2-phenyl-2-[(2RS)-piperidin-2-yl]ethanamide, (2RS)-2-phenyl-2-[(2RS)-piperidin-2-yl]acetamide, Methylphenidate Hydrochloride Imp. C (EP)
IUPAC-namn (2R)-2-phenyl-2-[(2R)-piperidin-2-yl]acetamide
LEDER NC(=O)[C@@H]([C@H]1CCCCN1)c2ccccc2
Specifikationer
Kemiskt namn eller material (2RS)-2-Phenyl-2-[(2RS)-piperidin-2-yl]acetamide
InChI formel InChI=1S/C13H18N2O/c14-13(16)12(10-6-2-1-3-7-10)11-8-4-5-9-15-11/h1-3,6-7,11-12,15H,4-5,8-9H2,(H2,14,16)/t11-,12-/m1/s1
Kvantitet 25 mg
Formel vikt 218.1419