Läs mer
D-Leucine Methyl Ester Hydrochloride, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | D-Leucine Methyl Ester Hydrochloride |
| CAS | 5845-53-4 |
| Molekylformel | C7 H15 N O2 . Cl H |
| Kvantitet | 1 g |
| Synonym | D-Leucine, methyl ester, hydrochloride (1:1), D-Leucine, methyl ester, hydrochloride (9CI), Leucine, methyl ester, hydrochloride, D- (8CI), Methyl D-leucinate hydrochloride, NSC 522846, (R)-Leucine methyl ester hydrochloride, (R)-Methyl 2-amino-4-methylpentanoate hydrochloride, O-Methyl-D-leucine hydrochloride, (R)-2-Amino-4-methylpentanoic acid methyl ester hydrochloride, D-Leucine methyl ester hydrochloride, Methyl (2R)-2-amino-4-methylpentanoate hydrochloride |
| InChI formel | InChI=1S/C7H15NO2.ClH/c1-5(2)4-6(8)7(9)10-3;/h5-6H,4,8H2,1-3H3;1H/t6-;/m1./s1 |
| LEDER | Cl.COC(=O)[C@H](N)CC(C)C |
| IUPAC-namn | methyl (2R)-2-amino-4-methylpentanoate;hydrochloride |
| Molekylvikt (g/mol) | 181.66 |
| Formel vikt | 181.087 |