Läs mer
MedChemExpress D-Luciferin potassium
Specifikationer
Specifikationer
| Kvantitet | 50 mg |
| Förvaringskrav | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Löslighet | H2O : 25 mg/mL (78.52 mM; Need ultrasonic) |
| Frakt skick | Room temperature |
| CAS | 115144-35-9 |
| Molekylvikt (g/mol) | 318.41 g/mol |
| Synonym | D-(-)-Luciferin potassium ∣ Firefly luciferin potassium ∣ Beetle Luciferin potassium |
| Mål | Dopamine Receptor |