Läs mer
D-Lyxonic Acid, Potassium Salt, TRC
Produktkod. 30465276
Change view
Kvantitet:
250 mg
1 g
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30465276 250 mg
30464532 1 g
30482115 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30465276 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL489950250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 78138-87-1
Molekylformel C5H9KO6
Molekylvikt (g/mol) 204.22
IUPAC-namn potassium;(2S,3S,4R)-2,3,4,5-tetrahydroxypentanoate
LEDER [K+].OC[C@@H](O)[C@H](O)[C@H](O)C(=O)[O-]
Specifikationer
Kemiskt namn eller material D-Lyxonic Acid, Potassium Salt
Kvantitet 250 mg
InChI formel InChI=1S/C5H10O6.K/c6-1-2(7)3(8)4(9)5(10)11;/h2-4,6-9H,1H2,(H,10,11);/q;+1/p-1/t2-,3+,4+;/m1./s1
Formel vikt 204.0036